Learn More
Medchemexpress LLC RBPJ Inhibitor-1 (RIN1) | 2682114-39-0 | 99.9% | 311.31 | 50 MG
SDP
Catalog No. 5000350822
Encompass_Preferred
RBPJ Inhibitor-1 (RIN1) is the first RBPJ inhibitor, which blocks the functional interaction of RBPJ with SHARP. It inhibits NOTCH-dependent tumor cell proliferation and is intended for research use only.
  • Blocks the functional interaction of RBPJ with SHARP
  • Inhibits NOTCH-dependent tumor cell proliferation
  • Inhibits the proliferation of hematologic cancer cell lines
  • Promotes skeletal muscle differentiation from C2C12 myoblasts
Catalog No. 50-003-50822 Supplier Medchemexpress LLC Supplier No. HY13747150MG
RBPJ Inhibitor-1 (RIN1) is the first RBPJ inhibitor, which blocks the functional interaction of RBPJ with SHARP. It inhibits NOTCH-dependent tumor cell proliferation and is intended for research use only.
  • Blocks the functional interaction of RBPJ with SHARP
  • Inhibits NOTCH-dependent tumor cell proliferation
  • Inhibits the proliferation of hematologic cancer cell lines
  • Promotes skeletal muscle differentiation from C2C12 myoblasts