Learn More
Aviva Systems Biology Corp RBPJL Antibody - N-terminal region (ARP34313_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34313_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBPJL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBPJL. Alternate names: SUH, SUHL, RBP-L, RBPSUHLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55340