Learn More
Aviva Systems Biology Corp RBPJL Antibody - middle region (ARP34710_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34710_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBPJL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBPJL. Alternate names: RBP-L, RBPSUHL, SUH, SUHLApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55526