Learn More
Aviva Systems Biology Corp RBPMS Antibody - N-terminal region (ARP40269_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40269_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBPMS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RBPMS. Alternate names: HERMESApplicationIHC, WB, ICC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57370