Learn More
Cayman Chemical (R)-Bromoenol lactone, Purity: ≥98%, CAS: 478288-90-3, 5 mg
SDP
Catalog No. 502888774
Encompass_Preferred
A selective iPLA2γ inhibitor (IC50 = ~0.6 µM); selective for iPLA2γ over iPLA2β at 20-30 µM
Catalog No. 50-288-8774 Supplier Cayman Chemical Supplier No. 100068005 MG
A selective iPLA2γ inhibitor (IC50 = ~0.6 µM); selective for iPLA2γ over iPLA2β at 20-30 µM