Learn More
Columbia Biosciences Corp RBT ANTI-V5 DYLIGHT 650 100UG
Supplier:  Columbia Biosciences Corp D101834
Encompass_Preferred
Rabbit anti-V5 tag IgG, primary antibody, conjugated to DyLight 650, 100ug for Flow Cytometry and Cell Based Assays
Catalog No. 50-815-76