Learn More
Innovative Research Inc Rbt Fbrnctn Ply Frctn Btn lbl
Supplier:  Innovative Research Inc IRBAMSFBNGFBL1MG
Encompass_Preferred
Rabbit Anti Mouse Fibronectin Polyclonal Fractionated Biotin Labeled 1mg - Rabbit Anti Mouse Fibronectin Polyclonal Fractionated Biotin Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using immobilized protein A/G chromatography followed by biotin labeling. Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted). This product can be used in plate-based assay analytics such as ELISA. Host Rabbit, Species Mouse, Target Fibronectin, Clonality Polyclonal, Purity IgG Fractionated Biotin Labeled, Source Rabbit Serum, Storage Conditions -70C. For Research Use Only.
Catalog No. 50-274-4064