Learn More
Innovative Research Inc Rbt IgG Frctn Pur 1gm
Supplier:  Innovative Research Inc IRBIGGGF1GM
Encompass_Preferred
Rabbit IgG Fractionated Purified 1gm - Rabbit IgG Fractionated Purified from Innovative Research has been purified from rabbit serum by fractionation. This is a frozen liquid buffered in 10mM sodium phosphate, 150mM sodium chloride pH 7.2 and a purity of greater than 97% as determined by SDS-PAGE analysis. This product is useful as a co-precipitant in radioimmunoassay methods that use polyethylene glycol or ammonium sulfate precipitaiton, and as a protein stabilizer blocker for immunoassays. This is also useful as an IgG control for biochemical analytical testing. Immunoglobulin G (IgG) is an antibody that plays a role in controlling infection, neutralizing toxins, activating the classical pathway of the complement system. Each individual IgG contains two identical antigen binding sites. There are four subclasses of IgG (IgG1, IgG2, IgG3, and IgG4) that have different (and sometimes opposing) properties.Storage Conditions -20C
Catalog No. 50-274-4046