Learn More
Innovative Research Inc Rbt IgG Frctn Pur Lyo 1gm
Catalog No. 502744045
Encompass_Preferred
Rabbit IgG Fractionated Purified Lyophilized 1gm - Rabbit IgG Fractionated Purified Lyophilized from Innovative Research has been purified from rabbit serum by fractionation. This is a lyophilized powder buffered in 10mM sodium phosphate, 150mM sodium chloride pH 7.2, with a purity of greater than 97% as determined by SDS-PAGE analysis. Immunoglobulin G (IgG) is an antibody that plays a role in controlling infection, neutralizing toxins, activating the classical pathway of the complement system, and more. Each individual IgG contains two identical antigen binding sites. There are four subclasses of IgG (IgG1, IgG2, IgG3, and IgG4) that have different (and sometimes opposing) properties.
Catalog No. 50-274-4045 Supplier Innovative Research Inc Supplier No. IRBIGGGFLY1GM
Rabbit IgG Fractionated Purified Lyophilized 1gm - Rabbit IgG Fractionated Purified Lyophilized from Innovative Research has been purified from rabbit serum by fractionation. This is a lyophilized powder buffered in 10mM sodium phosphate, 150mM sodium chloride pH 7.2, with a purity of greater than 97% as determined by SDS-PAGE analysis. Immunoglobulin G (IgG) is an antibody that plays a role in controlling infection, neutralizing toxins, activating the classical pathway of the complement system, and more. Each individual IgG contains two identical antigen binding sites. There are four subclasses of IgG (IgG1, IgG2, IgG3, and IgG4) that have different (and sometimes opposing) properties.