Learn More
Innovative Research Inc Rbt PAI1 Ply Frctn Btn lbl 1mg
Catalog No. 502744059
Encompass_Preferred
Rabbit Anti Mouse PAI-1 Polyclonal Fractionated Biotin Labeled 1mg - Rabbit Anti Mouse PAI-1 Polyclonal Fractionated Biotin Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antiserum is whole serum containing polyclonal antibodies (predominantly IgG, IgM) directed against the specified antigen and includes host antibodies and immunoglobulins of all classes along with other serum proteins. This product can be used in analytic tools such as ELISA kits and Western blot.
Catalog No. 50-274-4059 Supplier Innovative Research Inc Supplier No. IRBAMSPAI1GFBC1MG
Rabbit Anti Mouse PAI-1 Polyclonal Fractionated Biotin Labeled 1mg - Rabbit Anti Mouse PAI-1 Polyclonal Fractionated Biotin Labeled from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antiserum is whole serum containing polyclonal antibodies (predominantly IgG, IgM) directed against the specified antigen and includes host antibodies and immunoglobulins of all classes along with other serum proteins. This product can be used in analytic tools such as ELISA kits and Western blot.