Learn More
Innovative Research Inc Rbt tPA Ply Aff Pur 100ug
Catalog No. 502744072
Encompass_Preferred
Rabbit Anti Mouse/Rat tPA Polyclonal Affinity Purified 100ug - Rabbit Anti Mouse/Rat tPA Polyclonal Affinity Purified from Innovative Research is a polyclonal antibody in a Frozen liquid format buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. The affinity isolated antigen-specific antibody was purified from whole antiserum by immunospecific purification using antigen-bound agarose. This process removes the host serum proteins (including immunoglobulins) that do not specifically bind to the target antigen. This antibody preparation has the lowest amount of non-specific binding and therefore the highest titer. This product can be used in analytic tools such as ELISA, Capture, Western blot, Flow cytometry, Immunohistochemistry of frozen sections, Immunohistochemistry of paraffin sections, Immunohistochemistry of PFA fixed sections, Immunofluorescence, Immunoprecipitation and Affinity purification.
Catalog No. 50-274-4072 Supplier Innovative Research Inc Supplier No. IRBAMSAP100UG
Rabbit Anti Mouse/Rat tPA Polyclonal Affinity Purified 100ug - Rabbit Anti Mouse/Rat tPA Polyclonal Affinity Purified from Innovative Research is a polyclonal antibody in a Frozen liquid format buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6. The affinity isolated antigen-specific antibody was purified from whole antiserum by immunospecific purification using antigen-bound agarose. This process removes the host serum proteins (including immunoglobulins) that do not specifically bind to the target antigen. This antibody preparation has the lowest amount of non-specific binding and therefore the highest titer. This product can be used in analytic tools such as ELISA, Capture, Western blot, Flow cytometry, Immunohistochemistry of frozen sections, Immunohistochemistry of paraffin sections, Immunohistochemistry of PFA fixed sections, Immunofluorescence, Immunoprecipitation and Affinity purification.