Learn More
Innovative Research Inc Rbt tPA Ply Frctn 200ug
Supplier:  Innovative Research Inc IRBAMSTPAGF200UG
Encompass_Preferred
Rabbit Anti Mouse/Rat tPA Polyclonal Fractionated 200ug - Rabbit Anti Mouse/Rat tPA Polyclonal Fractionated from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein A affinity chromatography . Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted). This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of frozen sections and Immunoprecipitation.
Catalog No. 50-274-4053