Learn More
Innovative Research Inc Rbt Vtrnctn Ply Frctn Btn lbl
Catalog No. 502744052
Encompass_Preferred
Rabbit Anti Mouse Vitronectin Polyclonal Fractionated Biotin Labeled 100ug - Rabbit Anti Mouse Vitronectin Polyclonal Antiserum from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antiserum is whole serum containing polyclonal antibodies (predominantly IgG & IgM) directed against the specified antigen and includes host antibodies and immunoglobulins of all classes along with other serum proteins. This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of paraffin sections and Immunofluorescence.
Catalog No. 50-274-4052 Supplier Innovative Research Inc Supplier No. IRBAMSVTNGFBC100UG
Rabbit Anti Mouse Vitronectin Polyclonal Fractionated Biotin Labeled 100ug - Rabbit Anti Mouse Vitronectin Polyclonal Antiserum from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antiserum is whole serum containing polyclonal antibodies (predominantly IgG & IgM) directed against the specified antigen and includes host antibodies and immunoglobulins of all classes along with other serum proteins. This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of paraffin sections and Immunofluorescence.