Learn More
Innovative Research Inc Rbt X Ms Atplsmn Ply Frctn 1mg
Catalog No. 502744071
Encompass_Preferred
Rabbit Anti Mouse Antiplasmin Polyclonal Fractionated 1mg - Rabbit Anti Mouse Antiplasmin Polyclonal Fractionated from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein A affinity chromatography . Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted). This product can be used in analytic tools such as ELISA kits and Western blot.
Catalog No. 50-274-4071 Supplier Innovative Research Inc Supplier No. IRBAMSAPLGF1MG
Rabbit Anti Mouse Antiplasmin Polyclonal Fractionated 1mg - Rabbit Anti Mouse Antiplasmin Polyclonal Fractionated from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein A affinity chromatography . Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted). This product can be used in analytic tools such as ELISA kits and Western blot.