Learn More
Innovative Research Inc Rbt X Ms PAI1 Ply Frctn 1mg
Catalog No. 502744061
Encompass_Preferred
Rabbit Anti Mouse PAI-1 Polyclonal Fractionated 1mg - Rabbit Anti Mouse PAI-1 Polyclonal Fractionated from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein A affinity chromatography . Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted). This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of frozen sections and Immunohistochemistry of paraffin sections
Catalog No. 50-274-4061 Supplier Innovative Research Inc Supplier No. IRBAMSPAI1GF1MG
Rabbit Anti Mouse PAI-1 Polyclonal Fractionated 1mg - Rabbit Anti Mouse PAI-1 Polyclonal Fractionated from Innovative Research is a polyclonal antibody in a Frozen liquid format. This antibody has been purified using protein A affinity chromatography . Fractionated preparations may be especially useful in research where high affinity is necessary, especially in cases where the target antigen is less abundant. IgG Fractionated antiserum can also contain very high affinity antibodies that are sometimes missing from affinity purified preparations (since they can bind so tightly to the affinity matrix that they are not always eluted). This product can be used in analytic tools such as ELISA, Western blot, Immunohistochemistry of frozen sections and Immunohistochemistry of paraffin sections