Learn More
Aviva Systems Biology Corp RBX1 Antibody - middle region (AVARP03042_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03042_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RBX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBX1. Alternate names: ROC1, RNF75, BA554C12.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62525