Learn More
Enzo Life Sciences Rbx1 (human), (recombinant) (His-tag) (25 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW03950025
Encompass_Preferred
Alternative name: Ring box protein 1, ROC1, Regulator of cullins 1, Ring finger protein 75. Purity: ≥98% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, pH 8.0, containing 50mM NaCl and 1mM DTT. Source: Produced in E. coli BL21 (λDE3). Long Term Storage: -80°C.
Catalog No. 50-201-2248