Learn More
CPC Scientific H-DPhe-Cys-Tyr-DTrp-Lys-Val-Cys-Trp-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110431
Encompass
The somatostatin analog. Re-188-RC-160 (radioisotope RheniuM-188-labeled RC-160) has been successfully used for the local/regional treatment of experimental breast cancer and other cancers.ONE-LETTER SEQUENCE: fCYwKVCW-NH2 (C2&C7 bridge)MOLECULAR FORMULA: C57H72N12O9S2MOLECULAR WEIGHT:1133.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [103222-11-3], SYNONYMS: RC-160, SanvarRESEARCH AREA: GastrointestinalREFERENCES: P. O. Zamora et al., Hybridoma, 16, 85 (1997)
Catalog No. 50-211-0431 Supplier CPC Scientific Supplier No. SOMA007A
The somatostatin analog. Re-188-RC-160 (radioisotope RheniuM-188-labeled RC-160) has been successfully used for the local/regional treatment of experimental breast cancer and other cancers.ONE-LETTER SEQUENCE: fCYwKVCW-NH2 (C2&C7 bridge)MOLECULAR FORMULA: C57H72N12O9S2MOLECULAR WEIGHT:1133.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [103222-11-3], SYNONYMS: RC-160, SanvarRESEARCH AREA: GastrointestinalREFERENCES: P. O. Zamora et al., Hybridoma, 16, 85 (1997)