Learn More
Bio Rad Laboratories RC, DC PROTEIN
Catalog No. NC9876114
Encompass
RC DC Protein Assay Kit II, includes RC reagents package,DC protein assay reagents package,bovine serum albumin standard,500 std assays, a colorimetric assay for protein determination in the presence of reducing agents and detergents
Catalog No. NC9876114 Supplier Bio Rad Laboratories Supplier No. 5000122
RC DC Protein Assay Kit II, includes RC reagents package,DC protein assay reagents package,bovine serum albumin standard,500 std assays, a colorimetric assay for protein determination in the presence of reducing agents and detergents