Learn More
Aviva Systems Biology Corp RC3H2 Antibody - middle region (ARP43260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RC3H2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RC3H2. Alternate names: FLJ20301, FLJ20713, MGC52176, MNAB, RNF164ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58476