Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VACUUBRAND™ RC6 Chemistry-HYBRID Vacuum Pump

Catalog No. 13689411 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Plug Type:
US
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type
13-689-411 US
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-689-411 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20698563

missing translation for 'validMainframeMsg'

For fine-vacuum applications

Combines rotary vane and diaphragm vacuum pump technology to support freeze drying and molecular distillations, with substantially reduced maintenance and less hazardous oil waste. Chemistry-design diaphragm pump is used to back the rotary vane pump. This configuration keeps pump oil under vacuum, reducing condensation of solvent vapors in pump oil and oil changes by as much as 90% over conventional rotary vane designs.

  • Free air capacity: 4.1cfm
  • Ultimate vacuum: 4 x 10-4mbar
  • Restriction-free, high-flow inlet ensures high flowrates under vacuum
  • Includes integrated oil mist filter

Order Info

Dimensions: 20L x 12W x9 in. H (51 x 30.5 x 23cm). Net weight: 53 lb. (24.2kg).

TRUSTED_SUSTAINABILITY

Specifications

Max. Vacuum 0.75 x 10-2 torr (with Gas Ballast), 1.5 x 10-3 torr (without Gas Ballast)
Horsepower 0.5 hp
Amperage 5.1/5.7 A
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010
Electrical Requirements 100/120 V 50/60 Hz
Model RC6
Noise Level 50 dbA
Plug Type US
Weight (English) 53.35 lb.
Weight (Metric) 24.2 kg
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 12°C
Flow Rate 3.5/4.1 CFM
Frequency 50/60 Hz
Overall Height (English) 9.05 in.
Overall Height (Metric) 231.14 mm
Overall Length (English) 20 in.
Overall Length (Metric) 510.54 mm
Overall Width (English) 12 in.
Overall Width (Metric) 304.8 mm
Pump Type Chemistry-HYBRID Rotary Vane Pump
Enclosure Rating IP40
Resistance Properties Aggressive Gas-Resistant, Corrosion-Resistant
Temperature (English) Min 54°F
Oil Capacity 0.36 to 0.56 qt.
Oil Type Oil-Sealed
No. of Stages 4
Maximum Pressure 16 psi
Voltage 100/120 V
Storage Requirements -10°C to +60°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.