Learn More
Vector Laboratories Ricinus Communis Agglutinin I (RCA I, RCA120), Agarose bound
SDP
Catalog No. NC2745155
Encompass
Ricinus Communis Agglutinin I (RCA I, RCA120), Agarose bound (AL-1083-5). Agarose bound Ricinus communis agglutinin I is prepared using our affinity-purified lectins. Bead diameter ranges in size from 45-165 microns. Product supplied as a 1:1 suspension in buffer
Catalog No. NC2745155 Supplier Vector Laboratories Supplier No. AL10835
Ricinus Communis Agglutinin I (RCA I, RCA120), Agarose bound (AL-1083-5). Agarose bound Ricinus communis agglutinin I is prepared using our affinity-purified lectins. Bead diameter ranges in size from 45-165 microns. Product supplied as a 1:1 suspension in buffer