Learn More
Aviva Systems Biology Corp RCAN1 Antibody - middle region (ARP38457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCAN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RCAN1. Alternate names: ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53105