Learn More
Aviva Systems Biology Corp RCAN2 Antibody - middle region (ARP52109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCAN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RCAN2. Alternate names: CSP2, DSCR1L1, MCIP2, RCN2, ZAKI-4, ZAKI4, hRCN2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-637