Learn More
Aviva Systems Biology Corp RCC1 Antibody - C-terminal region (ARP61300_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61300_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCC1. It was validated on Western Blot. . Alternate names: CHC1, RCC1-I, SNHG3-RCC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-336