Learn More
Aviva Systems Biology Corp RCE1 Antibody - N-terminal region (ARP44797_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44797_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RCE1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RCE1. Alternate names: FACE2, RCE1A, RCE1BApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58895