Learn More
Medchemexpress LLC RCGD423 200mg | 108237-91-8 | 331.23 g/mol | 200 MG
SDP
Catalog No. 5000229713
Encompass_Preferred
RCGD423 is a small-molecule gp130 modulator used in research to prevent articular cartilage degeneration and promote repair. It modulates gp130 signaling and activates downstream JAK/STAT pathways, and has been applied in cellular and animal studies to evaluate cartilage protection and repair mechanisms.
  • Modulates gp130 signaling and activates JAK/STAT pathways.
  • Demonstrated disease-modifying activity in cartilage injury and degeneration models.
  • High purity listed (99.92%), suitable for biochemical and cellular assays.
  • Chemical formula C15H11BrN2S; molecular weight 331.23 g/mol.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-29713 Supplier Medchemexpress LLC Supplier No. HY114775200MG
RCGD423 is a small-molecule gp130 modulator used in research to prevent articular cartilage degeneration and promote repair. It modulates gp130 signaling and activates downstream JAK/STAT pathways, and has been applied in cellular and animal studies to evaluate cartilage protection and repair mechanisms.
  • Modulates gp130 signaling and activates JAK/STAT pathways.
  • Demonstrated disease-modifying activity in cartilage injury and degeneration models.
  • High purity listed (99.92%), suitable for biochemical and cellular assays.
  • Chemical formula C15H11BrN2S; molecular weight 331.23 g/mol.
  • For research use only; not for human or clinical use.