Learn More
Lonza Walkersville RCGM Rat Cardiomyocyte Growth
Supplier:  Lonza Walkersville CC4516
Encompass
RCGM Rat Cardiomyocyte Growth Medium is specifically formulated for the growth and survival of rat neonatal cardiac myocytes in culture Lonza offers customers a fully validated and optimized cardiomyocyte medium and growth factors to complement our Clonetics Rat Cardiomyocytes R-CM-561 as a complete system to study cardiac biology/toxicology Basal medium and SingleQuots Kit growth factors are available separately Media products are specially designed to support the growth of human and animal primary cells Media systems have been specifically developed and optimized for various cell systems Media are supplied in several formats Basal Medium - contains no growth factors cytokines or supplements SingleQuots - growth factors cytokines and supplements BulletKits - basal media and SingleQuots Kit packaged together
Catalog No. 50-004-96405