Learn More
Cole-Parmer Energizer NH15BP-2 Eveready Rechargeable NiMH Batteries, Size AA; Pack of 2
Catalog No. NC9375073 Shop All Cole-Parmer Products
Encompass_Preferred
Nickel Metal Hydride (NiMH) batteries provide 70 percent more power than NiCd batteries. They are an ideal alternative to disposable alkaline batteries. The batteries can be recharged and reused up to 1,000 times.
Catalog No. NC9375073 Supplier Cole-Parmer Supplier No. 0950641
Nickel Metal Hydride (NiMH) batteries provide 70 percent more power than NiCd batteries. They are an ideal alternative to disposable alkaline batteries. The batteries can be recharged and reused up to 1,000 times.