Learn More
Electron Microscopy Sciences EMS Rochester Pean Forceps with Curved, 9"
SDP
Catalog No. 501931480
Encompass_Preferred
These forceps feature a finger ring grip and adjustable interlocking cross arms for tight clamping of tissue. They have serrated tips made of stainless steel.
Tips: Curved
OAL: 9"
Catalog No. 50-193-1480 Supplier Electron Microscopy Sciences Supplier No. 7299911
These forceps feature a finger ring grip and adjustable interlocking cross arms for tight clamping of tissue. They have serrated tips made of stainless steel.
Tips: Curved
OAL: 9"