Learn More
Selleck Chemical LLC RCM-1 5mg
SDP
Supplier:  Selleck Chemical LLC S68985MG
Encompass_Preferred
RCM-1 is a nontoxic inhibitor of Forkhead box M1 (FOXM1) that suppresses goblet cell metaplasia and prevents IL-13 and STAT6 signaling in allergen-exposed mice. RCM-1 decreases carcinogenesis and nuclear ?-catenin. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3233