Learn More
Waters Corp Waters Corporation RCM Cartridge Holder, 8x10, for Radial-Pak Columns
Catalog No. 50818723 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The RCM 8x10 Cartridge Holder enhances efficiency by eliminating wall effect and voiding common to rigid columns. It works with 100 mm Radial-Pak Column segments, integrating into existing liquid chromatography systems without external utilities or column end-fittings.
  • Enhances efficiency by eliminating wall effect and voiding.
  • Compatible with 100 mm Radial-Pak Column segments.
  • Integrates into existing liquid chromatography systems without external utilities.
  • Radial compression module (RCM) applies pressure to flexible-wall columns.
  • Reduces wall effects, eliminates bubbles, voids, and channels for denser packing.
  • Achieves faster flow rates and improved resolution.
  • Manufactured in a cGMP, ISO 9001:2000 certified facility.
  • Utilizes ultra-pure reagents.
  • Each packing material batch undergoes stringent testing for quality.
Catalog No. 50-818-723 Supplier Waters Corp Supplier No. WAT082887
The RCM 8x10 Cartridge Holder enhances efficiency by eliminating wall effect and voiding common to rigid columns. It works with 100 mm Radial-Pak Column segments, integrating into existing liquid chromatography systems without external utilities or column end-fittings.
  • Enhances efficiency by eliminating wall effect and voiding.
  • Compatible with 100 mm Radial-Pak Column segments.
  • Integrates into existing liquid chromatography systems without external utilities.
  • Radial compression module (RCM) applies pressure to flexible-wall columns.
  • Reduces wall effects, eliminates bubbles, voids, and channels for denser packing.
  • Achieves faster flow rates and improved resolution.
  • Manufactured in a cGMP, ISO 9001:2000 certified facility.
  • Utilizes ultra-pure reagents.
  • Each packing material batch undergoes stringent testing for quality.