Learn More
Abcam Anti-Iba1 antibody, EPR16589, Rat IgG2a, Recombinant Monoclonal, 100 μL, Detects Iba1, Microglial marker, Human, Mouse, Rat reactivity, Applications: WB, IP, IHC-P, ICC/IF, Flow Cytometry
Supplier:  Abcam AB283346100UL
Encompass_Preferred
The Abcam Anti-Iba1 antibody [EPR16589] is a recombinant monoclonal antibody specifically designed for the detection of Allograft Inflammatory Factor 1 (Iba1), which is a well-established microglia marker validated in human, mouse, and rat samples. This antibody exhibits high batch-to-batch consistency, ensuring specificity and reliability across various applications, making it suitable for researchers in neurobiology and related fields. Its recombinant nature allows for better performance in immunodetection techniques.
Catalog No. NC2330816