Learn More
Abcam Recombinant Human CDKL5 protein, 20UG
Supplier:  Abcam AB13169520UG
Encompass_Preferred
Recombinant Human CDKL5 protein is a Human Fragment protein, in the 1 to 498 aa range, expressed in Baculovirus infected Sf9, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: STK9, CDKL5, Cyclin-dependent kinase-like 5, Serine/threonine-protein kinase 9
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1773444