Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creative Biomart Recombinant human USP19 protein, GST-tagged, 100 μg, 1-1290 a.a., deubiquitylase activity, stable at -70°C

Catalog No. NC3391515
Encompass

This recombinant protein, USP19, is an N-terminally Glutathione S-transferase (GST)-tagged enzyme produced in *E. coli*. It demonstrates notable deubiquitylase activity, effectively cleaving fluorogenic ubiquitin substrates. This protein is essential for studying muscle atrophy and oncology due to its role in regulating p27 degradation. USP19 stabilizes the E3 ubiquitin ligase RNF123, influencing muscle protein transcription and involvement in critical cellular processes during the G1 phase of the cell cycle.

  • Product type: Recombinant protein
  • Species: Human
  • Source: *E. coli*
  • Tag: N-terminal GST
  • Protein length: 1-1290 a.a.
  • Activity: Deubiquitylase enzyme activity
  • Storage stability: 12 months at -70°C
  • Biological function: Modulates myofibrillar protein transcription, regulates p27 levels
  • Research area: Muscle atrophy, oncology

Catalog No. NC3391515 Supplier Creative Biomart Supplier No. USP19186HCBMRS12102401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This recombinant protein, USP19, is an N-terminally Glutathione S-transferase (GST)-tagged enzyme produced in *E. coli*. It demonstrates notable deubiquitylase activity, effectively cleaving fluorogenic ubiquitin substrates. This protein is essential for studying muscle atrophy and oncology due to its role in regulating p27 degradation. USP19 stabilizes the E3 ubiquitin ligase RNF123, influencing muscle protein transcription and involvement in critical cellular processes during the G1 phase of the cell cycle.

  • Product type: Recombinant protein
  • Species: Human
  • Source: *E. coli*
  • Tag: N-terminal GST
  • Protein length: 1-1290 a.a.
  • Activity: Deubiquitylase enzyme activity
  • Storage stability: 12 months at -70°C
  • Biological function: Modulates myofibrillar protein transcription, regulates p27 levels
  • Research area: Muscle atrophy, oncology

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.