Learn More
Aviva Systems Biology Corp RCOR1 Antibody - C-terminal region (ARP39179_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39179_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCOR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RCOR1. Alternate names: RCOR, CORESTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse
Catalog No. 50-014-56931