Learn More
Aviva Systems Biology Corp RCOR1 Antibody - middle region (ARP34346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCOR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RCOR1. Alternate names: COREST, KIAA0071, RCORApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55350