Learn More
Aviva Systems Biology Corp RCOR3 Antibody - N-terminal region (ARP34132_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34132_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RCOR3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RCOR3. Alternate names: RP11-318L16.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55262