Learn More
Aviva Systems Biology Corp RCOR3 Antibody - N-terminal region (ARP34131_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34131_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RCOR3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RCOR3. Alternate names: FLJ10876, FLJ16298, RP11-318L16.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55261