Learn More
Cayman Chemical RCS-4 2-methoxy isomer, Purity: ≥98%, CAS: 1345966-76-8, 5 mg
SDP
Catalog No. 502951372
Encompass_Preferred
An analog of RCS-4 that differs only in the location of the methoxy group on the phenyl ring; structurally resembles JWH 250, which shows a high-affinity for both CB1 and CB2 (Ki = 11 and 33 nM, respectively)
Catalog No. 50-295-1372 Supplier Cayman Chemical Supplier No. 108655 MG
An analog of RCS-4 that differs only in the location of the methoxy group on the phenyl ring; structurally resembles JWH 250, which shows a high-affinity for both CB1 and CB2 (Ki = 11 and 33 nM, respectively)