Learn More
Cayman Chemical RCS-4 3-methoxy isomer, Purity: ≥98%, CAS: 1379922-51-6, 5 mg
SDP
Catalog No. 502951287
Encompass_Preferred
An RCS-4 analog whose design is based on the structure of the synthetic CB JWH 018 whose biological activity has not been reported; intended for forensic and research applications
Catalog No. 50-295-1287 Supplier Cayman Chemical Supplier No. 108665 MG
An RCS-4 analog whose design is based on the structure of the synthetic CB JWH 018 whose biological activity has not been reported; intended for forensic and research applications