Learn More
Cayman Chemical RCS-4-d9 (exempt preparation), Purity: ≥99% deuterated forms (d1-d9), CAS: 2749316-95-6, 1 mg
SDP
Catalog No. 502994881
Encompass_Preferred
An internal standard for the quantification of RCS-4 by GC- or LC-MS; provided as a DEA exempt preparation; intended for research and forensic applications
Catalog No. 50-299-4881 Supplier Cayman Chemical Supplier No. 228671 MG
An internal standard for the quantification of RCS-4 by GC- or LC-MS; provided as a DEA exempt preparation; intended for research and forensic applications