Learn More
Cayman Chemical RCS-4 M10 metabolite, Purity: ≥95%, CAS: 1952336-77-4, 1 mg
SDP
Catalog No. 502952885
Encompass_Preferred
A major urinary metabolite of RCS-4 that is potentially useful as a metabolic marker for the identification of RCS-4 ingestion; therefore intended for research and forensic purposes
Catalog No. 50-295-2885 Supplier Cayman Chemical Supplier No. 119781 MG
A major urinary metabolite of RCS-4 that is potentially useful as a metabolic marker for the identification of RCS-4 ingestion; therefore intended for research and forensic purposes