Learn More
Cayman Chemical RCS-4 M9 metabolite, Purity: ≥98%, CAS: 1630022-96-6, 10 mg
SDP
Catalog No. 502954887
Encompass_Preferred
An expected metabolite of RCS-4, characterized by hydroxylation on both the alkyl and phenyl groups; intended for research and forensic purposes
Catalog No. 50-295-4887 Supplier Cayman Chemical Supplier No. 1197710 MG
An expected metabolite of RCS-4, characterized by hydroxylation on both the alkyl and phenyl groups; intended for research and forensic purposes