Learn More
Cayman Chemical RCS-4 M9 metabolIte 1mg
SDP
Catalog No. 502954922
Encompass_Preferred
An expected metabolite of RCS-4, characterized by hydroxylation on both the alkyl and phenyl groups; intended for research and forensic purposes
Catalog No. 50-295-4922 Supplier Cayman Chemical Supplier No. 119771 MG
An expected metabolite of RCS-4, characterized by hydroxylation on both the alkyl and phenyl groups; intended for research and forensic purposes