Learn More
Cayman Chemical RCS-4 M9 metabolite, Purity: ≥98%, CAS: 1630022-96-6, 5 mg
SDP
Catalog No. 502955640
Encompass_Preferred
An expected metabolite of RCS-4, characterized by hydroxylation on both the alkyl and phenyl groups; intended for research and forensic purposes
Catalog No. 50-295-5640 Supplier Cayman Chemical Supplier No. 119775 MG
An expected metabolite of RCS-4, characterized by hydroxylation on both the alkyl and phenyl groups; intended for research and forensic purposes