Learn More
Cayman Chemical RCS-8 3-methoxy isomer, Purity: ≥98%, CAS: 1427326-08-6, 10 mg
SDP
Catalog No. 502951510
Encompass_Preferred
Differs from RCS-8 by having a methoxy group at the 3, rather than 2, position of its phenylacetyl group; intended for forensic and research applications
Catalog No. 50-295-1510 Supplier Cayman Chemical Supplier No. 1086410 MG
Differs from RCS-8 by having a methoxy group at the 3, rather than 2, position of its phenylacetyl group; intended for forensic and research applications