Learn More
Cayman Chemical RCS-8 4-methoxy isomer, Purity: ≥95%, CAS: 1427326-10-0, 1 mg
SDP
Catalog No. 502951630
Encompass_Preferred
Differs from RCS-8 by having a methoxy group at the 4, rather than 2, position of its phenylacetyl group; intended for forensic and research applications
Catalog No. 50-295-1630 Supplier Cayman Chemical Supplier No. 108631 MG
Differs from RCS-8 by having a methoxy group at the 4, rather than 2, position of its phenylacetyl group; intended for forensic and research applications