Learn More
TA Instruments RCS Gas Dryer, Molecular Sieve Cartridge, Removes Trace Moisture in Purge Gases, Provides Dry Gas to-90°C Dew Point, Extends LNCS Dryer Cartridge Life, Compatible with LNCS Gas Dryer
Catalog No. 5000580824 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments RCS Gas Dryer is a molecular sieve cartridge dryer for removal of trace moisture in purge gases. It provides dry gas to -90°C dew point and can be used in conjunction with the LNCS gas dryer to extend the life of the LNCS dryer cartridge.
Catalog No. 50-005-80824 Supplier TA Instruments Supplier No. 200266.001
Waters TA Instruments RCS Gas Dryer is a molecular sieve cartridge dryer for removal of trace moisture in purge gases. It provides dry gas to -90°C dew point and can be used in conjunction with the LNCS gas dryer to extend the life of the LNCS dryer cartridge.