Learn More
Cayman Chemical RCS-4 4-hydroxyphenyl metabolite, Purity: ≥98%, CAS: 1443988-18-8, 10 mg
SDP
Catalog No. 502954002
Encompass_Preferred
An expected urinary metabolite of RCS-4 that is potentially useful as a metabolic marker for the identification of RCS-4 ingestion; intended for research and forensic purposes
Catalog No. 50-295-4002 Supplier Cayman Chemical Supplier No. 1434510 MG
An expected urinary metabolite of RCS-4 that is potentially useful as a metabolic marker for the identification of RCS-4 ingestion; intended for research and forensic purposes